siRNA Details
Gene Symbol: A430102A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020755.1 | - | 1 | 434 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724641.1 | - | 1 | 431 | Chr. 11 - 120932952 - 120933567 [+] on Build GRCm38 |
