siRNA Details
Gene Symbol: Gm20759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046030.1 | 1 | 65 | |||
GenBank mRNA | AK135737.1 | - | 1 | 66 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ052541.1 | - | 1 | 66 | Chr. 11 - 86002914 - 86024350 [-] on Build GRCm38 |
