siRNA Details
Gene Symbol: 4833419O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014730.1 | - | 1 | 92 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714228.1 | - | 1 | 88 | Chr. 16 - 20800465 - 20803382 [-] on Build GRCm38 |
