siRNA Details
Gene Symbol: 4930570B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016261.1 | - | 1 | 593 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715874.1 | - | 1 | 591 | Chr. 15 - 30598250 - 30626620 [-] on Build GRCm38 |
