siRNA Details
Gene Symbol: 4930570G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040399.1 | 4 | 665 | |||
GenBank mRNA | AK016265.1 | - | 4 | 665 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715880.1 | - | 4 | 665 | Chr. 3 - 156546848 - 156561687 [-] on Build GRCm38 |
