siRNA Details
Gene Symbol: 5830408B19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA268496.1 | - | 1 | 120 | ||
AK017909.1 | - | 1 | 1690 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM942884.1 | - | 1 | 35 | Chr. 10 - 9622945 - 9623184 [-] on Build GRCm38 |
