siRNA Details
Gene Symbol: 4930480M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046278.1 | 3 | 297 | |||
GenBank mRNA | AK015603.1 | - | 3 | 298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715155.1 | - | 3 | 294 | Chr. 12 - 26211270 - 26240707 [+] on Build GRCm38 |
