siRNA Details
Gene Symbol: Gm32441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131795.1 | - | 3 | 438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707336.1 | - | 3 | 438 | Chr. 19 - 56656511 - 56664611 [-] on Build GRCm38 |
