siRNA Details
Gene Symbol: 4930540A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016005.1 | - | 1 | 355 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715583.1 | - | 1 | 355 | Chr. 8 - 8831447 - 8831907 [-] on Build GRCm38 |
