siRNA Details
Gene Symbol: 4930539M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040597.1 | 2 | 278 | |||
GenBank mRNA | AK016002.1 | - | 2 | 285 | ||
BB615931.1 | - | 2 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715580.1 | - | 2 | 281 | Chr. 3 - 9061970 - 9071054 [+] on Build GRCm38 |
