siRNA Details
Gene Symbol: Gm20597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037973.1 | 4 | 3647 | |||
GenBank mRNA | AK045844.1 | - | 1 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB643618.1 | - | 1 | 388 | Chr. 10 - 53500015 - 53500669 [-] on Build GRCm38 |
