siRNA Details
Gene Symbol: 9530020I12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020565.1 | - | 3 | 295 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722031.1 | - | 3 | 295 | Chr. 12 - 11203086 - 11204409 [-] on Build GRCm38 |
