siRNA Details
Gene Symbol: 2510010E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010955.1 | - | 1 | 447 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710344.1 | - | 1 | 442 | Chr. 14 - 20545454 - 20546162 [-] on Build GRCm38 |
