siRNA Details
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054438.1 | - | 1, 2 | 163 | ||
BC150711.1 | - | 134 | 9087622 | |||
CJ056316.1 | - | 1, 2 | 227 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054438.1 | - | 1, 2 | 163 | ||
BC150711.1 | - | 134 | 9087622 | |||
CJ056316.1 | - | 1, 2 | 227 |