siRNA Details
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054438.1 | - | 4 | 736 | ||
BC150711.1 | - | 686 | 9087622 | |||
CJ065456.1 | - | 2 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB556821.2 | - | 3 | 562 | Chr. 1 - 173732282 - 173736457 [-] on Build GRCm38 |
