siRNA Details
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054438.1 | - | 1 | 107 | ||
AK138972.1 | - | 1 | 107 | |||
AK143243.1 | - | 1 | 95 | |||
BC150711.1 | - | 78 | 9087622 | |||
CJ056316.1 | - | 1 | 171 |
