siRNA Details
Gene Symbol: Gm36186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054439.1 | - | 2 | 1905 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB556827.2 | - | 2 | 508 | Chr. 5 - 148712862 - 148718397 [+] on Build GRCm38 |
