siRNA Details
Gene Symbol: 1700020L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006171.1 | - | 1 | 222 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706271.1 | - | 1 | 217 | Chr. 2 - 32082365 - 32082638 [+] on Build GRCm38 |
