siRNA Details
Gene Symbol: C330008A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021191.1 | - | 2 | 435 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728756.1 | - | 2 | 430 | Chr. 18 - 35913135 - 35916753 [+] on Build GRCm38 |
