siRNA Details
Gene Symbol: Gm36229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144827.1 | - | 3 | 252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734461.1 | - | 3 | 252 | Chr. 10 - 53409880 - 53423133 [+] on Build GRCm38 |
