siRNA Details
Gene Symbol: 1300014J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005005.2 | - | 1 | 245 | ||
AK040614.1 | - | 1 | 264 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB636312.1 | - | 1 | 264 | Chr. 12 - 80115863 - 80116503 [+] on Build GRCm38 |
