siRNA Details
Gene Symbol: 2310015D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037997.1 | 4 | 2426 | |||
GenBank mRNA | AK009355.1 | - | 1 | 1112 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310015D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037997.1 | 4 | 2426 | |||
GenBank mRNA | AK009355.1 | - | 1 | 1112 |