siRNA Details
Gene Symbol: 4930456K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015479.1 | - | 1 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715027.1 | - | 1 | 304 | Chr. 10 - 126693437 - 126694253 [+] on Build GRCm38 |
