siRNA Details
Gene Symbol: 4930405A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015475.1 | - | 1 | 19 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715024.1 | - | 1 | 19 | Chr. 14 - 22507276 - 22542798 [-] on Build GRCm38 |
