siRNA Details
Gene Symbol: 4930405A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046307.1 | 4 | 685 | |||
GenBank mRNA | AK015475.1 | - | 4 | 687 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715024.1 | - | 4 | 687 | Chr. 14 - 22507276 - 22542798 [-] on Build GRCm38 |
