siRNA Details
Gene Symbol: 3110023J12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014075.1 | - | 1 | 27 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713075.1 | - | 1 | 27 | Chr. 2 - 60232448 - 60232887 [+] on Build GRCm38 |
