siRNA Details
Gene Symbol: 4930433B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040544.1 | 1 | 125 | |||
GenBank mRNA | AK015298.1 | - | 1 | 126 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930433B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040544.1 | 1 | 125 | |||
GenBank mRNA | AK015298.1 | - | 1 | 126 |