siRNA Details
Gene Symbol: Gm32618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089548.1 | - | 600 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB232528.1 | - | 4 | 603 | Chr. 15 - 32778900 - 32795369 [+] on Build GRCm38 |
