siRNA Details
Gene Symbol: 4933426B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016924.1 | - | 1 | 353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716604.1 | - | 1 | 353 | Chr. 17 - 6808509 - 6818029 [+] on Build GRCm38 |
