siRNA Details
Gene Symbol: Prkag2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040684.1 | 3 | 425 | |||
GenBank mRNA | AK016926.1 | - | 3 | 424 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716606.1 | - | 421 | Chr. - - [-] on Build |
