siRNA Details
Gene Symbol: Gm13657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035219.1 | - | 4 | 1441 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB106723.2 | - | 1 | 166 | Chr. 2 - 75781642 - 75782306 [+] on Build GRCm38 |
