siRNA Details
Gene Symbol: Gm30146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082843.1 | - | 2 | 147 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728877.1 | - | 2 | 147 | Chr. 7 - 25860140 - 25869883 [+] on Build GRCm38 |
