siRNA Details
Gene Symbol: 1700054M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045919.1 | 3 | 185 | |||
GenBank mRNA | AK006785.1 | - | 3 | 185 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706771.1 | - | 3 | 185 | Chr. 2 - 118304914 - 118308166 [+] on Build GRCm38 |
