siRNA Details
Gene Symbol: Gm13427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006784.1 | - | 629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706770.1 | - | 4 | 629 | Chr. 2 - 31676374 - 31688469 [-] on Build GRCm38 |
