siRNA Details
Gene Symbol: Gm33677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132971.1 | - | 3 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614420.1 | - | 3 | 365 | Chr. 10 - 119030370 - 119050499 [-] on Build GRCm38 |
