siRNA Details
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013067.1 | - | 541 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712120.1 | - | 1 | 538 | Chr. 7 - 143549317 - 143549887 [+] on Build GRCm38 |
