siRNA Details
Gene Symbol: 2410021H03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045428.1 | 3 | 460 | |||
GenBank mRNA | AK010563.1 | - | 3 | 465 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709978.1 | - | 3 | 463 | Chr. 17 - 69275378 - 69277208 [-] on Build GRCm38 |
