siRNA Details
Gene Symbol: 4930589O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019834.1 | - | 881 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN691759.1 | - | 1 | 610 | Chr. 5 - 65493367 - 65494019 [+] on Build GRCm38 |
