siRNA Details
Gene Symbol: 4930593C16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040753.1 | 2, 3 | 347 | |||
GenBank mRNA | AK019838.1 | - | 2, 3 | 352 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930593C16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040753.1 | 2, 3 | 347 | |||
GenBank mRNA | AK019838.1 | - | 2, 3 | 352 |