siRNA Details
Gene Symbol: Gm34068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139592.1 | - | 3 | 481 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725418.1 | - | 3 | 481 | Chr. 1 - 183748977 - 183809749 [+] on Build GRCm38 |
