siRNA Details
Gene Symbol: Gm2516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046067.1 | 2 | 75 | |||
GenBank mRNA | AK135701.1 | - | 2 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719664.1 | - | 2 | 78 | Chr. 8 - 50368112 - 50397644 [+] on Build GRCm38 |
