siRNA Details
Gene Symbol: Gm2516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046067.1 | 5 | 659 | |||
GenBank mRNA | AK135701.1 | - | 5 | 663 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm2516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046067.1 | 5 | 659 | |||
GenBank mRNA | AK135701.1 | - | 5 | 663 |