siRNA Details
Gene Symbol: Gm30853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076946.1 | - | 6 | 730 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB017528.2 | - | 4 | 600 | Chr. 10 - 65875788 - 65898849 [-] on Build GRCm38 |
