siRNA Details
Gene Symbol: 4930455C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076940.1 | - | 1 | 577 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB017040.2 | - | 1 | 586 | Chr. 10 - 21341365 - 21342028 [-] on Build GRCm38 |
