siRNA Details
Gene Symbol: Gm16853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045742.1 | 1 | 165 | |||
GenBank mRNA | AK076941.1 | - | 1 | 167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615851.1 | - | 1 | 167 | Chr. 9 - 21406888 - 21411720 [-] on Build GRCm38 |
