siRNA Details
Gene Symbol: Snhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036616.1 | - | 1 | 335 | ||
AK051045.1 | - | 1 | 333 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U40654.1 | - | 1 | 490 | Chr. - - [-] on Build |
