siRNA Details
Gene Symbol: Gm33280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137601.1 | - | 213 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB629668.1 | - | 1 | 213 | Chr. 1 - 36132042 - 36133167 [+] on Build GRCm38 |
