siRNA Details
Gene Symbol: 4930479H17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019648.1 | - | 2 | 148 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715142.1 | - | 2 | 148 | Chr. 7 - 30347640 - 30353321 [-] on Build GRCm38 |
