siRNA Details
Gene Symbol: BC051077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132074.1 | - | 1 | 196 | ||
BC051077.1 | - | 1 | 89 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ933808.1 | - | 88 | Chr. - - [-] on Build |
