siRNA Details
Gene Symbol: Gm31169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156678.1 | - | 2 | 449 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY751489.1 | - | 2 | 449 | Chr. 6 - 143149390 - 143153931 [-] on Build GRCm38 |
